Anti-IFITM1 Rabbit Polyclonal Antibody
Catalog # 10206-680
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Interferon Induced Transmembrane Protein 1
- Antigen symbol:IFITM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Interferon-induced transmembrane protein 1|IFITM1|DSPA2a|Dispanin subfamily A member 2a|IFI17|9-27|CD225|LEU13
- Amino acid number:1 to 17
- Sequence:MHKEEHEVAVLGPPPST
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human IFITM1 (1-17aa MHKEEHEVAVLGPPPST).
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-680
- Supplier No.:PA1112
Specifications
About this item
Rabbit IgG polyclonal antibody for Interferon-induced transmembrane protein 1(IFITM1) detection. Tested with WB, IHC-P, IHC-F in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: IFITM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human