Anti-HSD11B1 Rabbit Polyclonal Antibody
Catalog # 10206-600
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Hydroxysteroid 11-beta Dehydrogenase 1
- Antigen symbol:HSD11B1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:corticosteroid 11-beta-dehydrogenase isozyme 1|HSD11B1|11-beta-hydroxysteroid dehydrogenase 1|HSD11L|HDL|MGC13539|short chain dehydrogenase/reductase family 26C|OTTHUMP00000034650|hydroxysteroid (11-beta) dehydrogenase 1|SDR26C1|HSD11B|member 1|11-DH|HSD11|11-beta-HSD1
- Amino acid number:16 to 33
- Sequence:MAYYYYSANEEFRPEMLQ
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human HSD11B1 (16-33aa MAYYYYSANEEFRPEMLQ), different from the related mouse sequence by two amino acids, and from the related rat sequence by four amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-600
- Supplier No.:PA1372-1
Specifications
About this item
Rabbit IgG polyclonal antibody for Corticosteroid 11-beta-dehydrogenase isozyme 1(HSD11B1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: HSD11B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat