Anti-CXCL1 Rabbit Polyclonal Antibody
Catalog # 10206-496
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C Motif) Ligand 1
- Antigen symbol:CXCL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:SCYB1|GROa|GRO1|MGSA-a|NAP-3|MGSA|FSP
- Amino acid number:99 to 107
- Sequence:KMLNSDKSN
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human GRO alpha (99-107aa KMLNSDKSN).
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-496
- Supplier No.:PA1760
Specifications
About this item
Rabbit IgG polyclonal antibody for Growth-regulated alpha protein(CXCL1) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CXCL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: