Anti-AIMP2/p38 Rabbit Polyclonal Antibody
Catalog # 10206-486
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:AIMP2/p38
- Antigen symbol:AIMP2/p38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:298 to 320
- Sequence:NVQRWMRSCENLAPFNTALKLLK
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human AIMP2/p38 (298-320aa NVQRWMRSCENLAPFNTALKLLK), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-486
- Supplier No.:PA1481
Specifications
About this item
Rabbit IgG polyclonal antibody for Aminoacyl tRNA synthase complex-interacting multifunctional protein 2(AIMP2) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: AIMP2/p38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat