Anti-CASP10 Rabbit Polyclonal Antibody
Catalog # 10206-408
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Caspase 10
- Antigen symbol:CASP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Caspase 10 apoptosis-related cysteine peptidase|FLICE2|ALPS2|MCH4
- Amino acid number:220 to 236
- Sequence:VKTFLEALPRAAVYRMN
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human CASP10 (220-236aa VKTFLEALPRAAVYRMN).
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-408
- Supplier No.:PA1513
Specifications
About this item
Rabbit IgG polyclonal antibody for Caspase-10(CASP10) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: CASP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human