Anti-GPR2 Rabbit Polyclonal Antibody
Catalog # 10206-404
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:G Protein-coupled Receptor 2
- Antigen symbol:GPR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:GPR2|C-C chemokine receptor type 10|G-protein coupled receptor 2|CC chemokine receptor 10|CC-CKR-10|G protein-coupled receptor 2|CCR-10|C-C CKR-10
- Amino acid number:312 to 327
- Sequence:FLGLRFRQDLRRLLRG
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human GPR2/CCR10 (312-327aa FLGLRFRQDLRRLLRG), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-404
- Supplier No.:PA1491
Specifications
About this item
Rabbit IgG polyclonal antibody for C-C chemokine receptor type 10(CCR10) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: GPR2/CCR10
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat