Anti-AQP4 Rabbit Polyclonal Antibody
10206-372
- Antibody type:Primary
- Antigen name:Aquaporin 4
- Antigen symbol:AQP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:WCH4|AQP-4|Aquaporin4|MIWC|Aquaporin-4|Mercurial-insensitive water channel
- Amino acid number:258 to 274
- Sequence:FKRRFKEAFSKAAQQTK
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Aquaporin 4 (258-274aa FKRRFKEAFSKAAQQTK), different from the related rat and mouse sequences by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-372
- Supplier No.:PA1937
Rabbit IgG polyclonal antibody for Aquaporin-4(AQP4) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: AQP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat