Anti-Caspase-1(P20) Rabbit Polyclonal Antibody
Catalog # 10206-318
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Caspase 1
- Antigen symbol:CASP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:N-terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:135 to 152
- Sequence:CSLEEAQRIWKQKSAEIY
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human CASP1 (135-152aa CSLEEAQRIWKQKSAEIY).
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-318
- Supplier No.:PA1440
Specifications
About this item
Rabbit IgG polyclonal antibody for Caspase-1(CASP1) detection. Tested with WB, IHC-P in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Caspase-1(P20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human