Anti-ARB1 Rabbit Polyclonal Antibody
Catalog # 10206-298
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Arrestin beta 1
- Antigen symbol:ARB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ARB1|C-arrestin |cArr|X-arrestin |ARR1|beta-arrestin-1|arrestin 2|CAR
- Amino acid number:82 to 103
- Sequence:ANVQSFPPAPEDKKPLTRLQER
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human beta Arrestin 1 (82-103aa ANVQSFPPAPEDKKPLTRLQER), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-298
- Supplier No.:PA2155
Specifications
About this item
Rabbit IgG polyclonal antibody for Beta-arrestin-1(ARRB1) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: ARB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat