Anti-PDPN Rabbit Polyclonal Antibody
Catalog # 10206-266
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Podoplanin
- Antigen symbol:PDPN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:T1A-2|36-KD|glycoprotein|PA2.26 antigen|hT1alpha-2|hT1alpha-1|AGGRUS|T1A|HT1A-1|GP36|Gp38|PA2.26|CTA-520D8.1|lung type-I cell membrane-associated glycoprotein (T1A-2)|T1-alpha|lung type I cell membrane associated glycoprotein|glycoprotein 36|OTS8|GP40
- Amino acid number:155 to 166
- Sequence:VVMRKISGRFSP
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of rat Podoplanin/gp36 (155-166aa VVMRKISGRFSP), different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-266
- Supplier No.:PA1675
Specifications
About this item
Rabbit IgG polyclonal antibody for Podoplanin(PDPN) detection. Tested with WB in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: PDPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat