Order Entry
Canada
ContactUsLinkComponent
Anti-Secretogranin 3 Rabbit Polyclonal Antibody
Anti-Secretogranin 3 Rabbit Polyclonal Antibody
Catalog # 10206-238
Anti-Secretogranin 3 Rabbit Polyclonal Antibody
Catalog # 10206-238
Supplier Number:  PA1071

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Secretogranin 3
  • Antigen symbol:
    SCG3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Amino acid number:
    438 to 450
  • Sequence:
    DFINKQADAYVEK
  • Storage temperature:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Secretogranin 3 (438-450aa DFINKQADAYVEK), different from the related mouse and rat sequences by one amino acid.
  • Purification:
    purified by immunogen affinity purification
  • Cat. no.:
    10206-238
  • Supplier No.:
    PA1071

Specifications

About this item

Rabbit IgG polyclonal antibody for Secretogranin-3(SCG3) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.

Reconstitute with distilled water

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Type: Primary
Antigen: Secretogranin 3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat