Anti-Bone Sialoprotein Rabbit Polyclonal Antibody
Catalog # 10206-180
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Bone Sialoprotein
- Antigen symbol:Bone Sialoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:17 to 34
- Sequence:FSMKNLHRRVKIEDSEEN
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human Bone Sialoprotein (17-34aa FSMKNLHRRVKIEDSEEN), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-180
- Supplier No.:PA1505
Specifications
About this item
Rabbit IgG polyclonal antibody for Bone sialoprotein 2(IBSP) detection. Tested with WB, IHC-P in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Bone Sialoprotein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat