Anti-NF68 Mouse Monoclonal Antibody [clone: NF-68]
10206-170
- Antibody type:Primary
- Antigen symbol:NF68
- Clonality:Monoclonal
- Clone:NF-68
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Rat
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Storage temperature:–20 °C
- Immunogen:Pig spinal cord.
- Cat. no.:10206-170
- Supplier No.:MA1070
Mouse IgG monoclonal antibody for NF68, neurofilament, light polypeptide (NEFL ) detection. Tested with WB, IHC-P, IHC-F in Human;pig;rat. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: NF68
Clonality: monoclonal
Clone: NF-68
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig, Rat