Anti-C Reactive Protein Rabbit Polyclonal Antibody
10206-132
- Antibody type:Primary
- Antigen name:C Reactive Protein
- Antigen symbol:C Reactive Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:209 to 224
- Sequence:KYEVQGEVFTKPQLWP
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human C Reactive Protein (209-224aa KYEVQGEVFTKPQLWP).
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-132
- Supplier No.:PA1028
Rabbit IgG polyclonal antibody for C-reactive protein(CRP) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: C Reactive Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human