Anti-DopamineR D5 Rabbit Polyclonal Antibody
Catalog # 10206-104
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:DopamineR D5
- Antigen symbol:DRD5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:388 to 407
- Sequence:LISYNQDTVFHREIAAAYVH
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse Dopamine Receptor D5 (388-407aa LISYNQDTVFHREIAAAYVH), different from the related rat sequence by two amino acids, and from the related human sequence by three amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10206-104
- Supplier No.:PA2204
Specifications
About this item
Rabbit IgG polyclonal antibody for D(1B) dopamine receptor(DRD5) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: DopamineR D5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat