Anti-CCNA1 Mouse Monoclonal Antibody [clone: CY-28]
Catalog # 10206-018
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Cyclin A
- Antigen symbol:CCNA1
- Clonality:Monoclonal
- Clone:CY-28
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Bovine,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilized
- Antigen synonyms:CCN1|CCNA
- Storage temperature:–20 °C
- Immunogen:Recombinant bovine cyclin A.
- Cat. no.:10206-018
- Supplier No.:MA1032
Specifications
About this item
Mouse IgG monoclonal antibody for Cyclin A, cyclin A2 (CCNA2) detection. Tested with WB, ICC in Human;mouse. No cross reactivity with other proteins.
Reconstitute with 1.2% sodium acetate or neutral PBS.
If 1ml of PBS is used, the antibody concentration will be 100ug/ml.
Type: Primary
Antigen: CCNA1
Clonality: Monoclonal
Clone: CY-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Bovine, Mouse