Anti-Glucose Transporter 5 GLUT5 Rabbit Polyclonal Antibody
Catalog # 10205-916
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Glucose Transporter 5
- Antigen symbol:GLUT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:232 to 251
- Sequence:ALQTLRGWKDVHLEMEEIRK
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of mouse Glucose Transporter 5 GLUT5 (232-251aa ALQTLRGWKDVHLEMEEIRK), different from the related rat sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10205-916
- Supplier No.:PA1737
Specifications
About this item
Rabbit IgG polyclonal antibody for Solute carrier family 2, facilitated glucose transporter member 5(SLC2A5) detection. Tested with WB, IHC-P, IHC-F in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Glucose Transporter 5 GLUT5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat