Anti-FABP5 Rabbit Polyclonal Antibody
Catalog # 10205-910
Supplier: Boster Biological Technology
Specifications
- Antibody type:Primary
- Antigen name:Fatty Acid Binding Protein 5 (Psoriasis-Associated)
- Antigen symbol:FABP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Epitope:near the N-terminus
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:EFABP|PA-FABP|E-FABP|PAFABP|KFABP
- Amino acid number:10 to 23
- Sequence:KWRLMESHGFEEYM
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of mouse Fatty Acid Binding Protein 5 (10-23aa KWRLMESHGFEEYM), different from the related rat sequence by two amino acids.
- Purification:purified by immunogen affinity purification
- Cat. no.:10205-910
- Supplier No.:PA1853
Specifications
About this item
Rabbit IgG polyclonal antibody for Fatty acid-binding protein, epidermal(FABP5) detection. Tested with WB, IHC-P in Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Fatty Acid Binding Protein 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat