Anti-Prostaglandin E Receptor EP1 Rabbit Polyclonal Antibody
10205-906
- Antibody type:Primary
- Antigen name:Prostaglandin E Receptor EP1
- Antigen symbol:PTGER1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:387 to 404
- Sequence:AWEASSLRSSRHSGLSHF
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Prostaglandin E Receptor EP1 (387-404aa AWEASSLRSSRHSGLSHF).
- Purification:purified by immunogen affinity purification
- Cat. no.:10205-906
- Supplier No.:PA1583
Rabbit IgG polyclonal antibody for Prostaglandin E2 receptor EP1 subtype(PTGER1) detection. Tested with WB in Human.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: Prostaglandin E Receptor EP1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human