Specifications
- Description:CarboFerm™ Neisseria Kit
- Cat. no.:CA10001-532
- Supplier No.:Z98
Specifications
About this item
CarboFerm I, rapid carbohydrate test for Neisseria and Moraxella, 24 tests per kit.
For the rapid identification and differentiation of Neisseria spp., including N. gonorrhoeae and N. meningitidis, and Moraxella (Branhamella) catarrhalis. CarboFerm™ uses acid production from carbohydrates to differentiate and identify Neisseria species and M. catarrhalis. Performs a glucose, lactose, maltose, sucrose, and butyrate test. CarboFerm™ Neisseria Kit reduces the time required from the usual 48 hours to as little as 2 - 4 hours!