ContactUsLinkComponent
Upchurch Scientific® Sample Loops for High Pressure Valves, IDEX Health & Science
  53501-296
Upchurch Scientific® Sample Loops for High Pressure Valves, IDEX Health & Science
  53501-296
 :  1876
  • Tubing ext.Ø:
    ⅛"
  • Tubing int.Ø:
    2.03 mm
  • Capacity:
    10000 µl
  • Description:
    Sample loop
  • Material:
    Stainless steel
  • Cat. no.:
    53501-296
  • For Use With:
    7125, 7010
  • Supplier No.:
    1876