SwabTool™ Swab Holder, Micronova
Catalog #: 10753-252
Supplier: Micronova
Small Business Enterprise
Specifications
- Description:SwabTool™ adapter
- Material:Stainless steel 304, electropolished, acetal components
- Supplier No.:QDSWT-01
Specifications
About this item
A custom tool specifically designed to hold swabs for environmental monitoring and sampling. Use with SSU series handles for extension in large tanks and other hard to reach areas.
- Swab is locked in place with a twist knob
- Adapted to swabs with a 6.4 mm (1/4 in) handle
- Optional 'straws' available to hold smaller or longer swabs
- Crafted from electropolished stainless steel with acetal components
- Quick Disconnect fitting connects to SSU series stainless steel handles
- Compatible with strong disinfectants
- Autoclavable
The Micronova SwabTool™ Adapter is compatible with a range of swabs when used with optional 'straws' to hold those which aresmaller or longer. Choose from the range of SSU series handles to match the length required for reach.
Customers Who Bought This Also Bought