ContactUsLinkComponent
Clute Patterns Gloves, Economy Leather Palm, Select Grade, MCR Safety
Clute Patterns Gloves, Economy Leather Palm, Select Grade, MCR Safety
Catalog # 80081-234
Supplier:  MCR Safety
undefined
Clute Patterns Gloves, Economy Leather Palm, Select Grade, MCR Safety
Catalog # 80081-234
Supplier:  MCR Safety
Supplier Number:  1040
Specifications
  • Glove Color:
    Gray,
    Red
  • Glove size:
    L
  • Ambidextrous/Hand-specific:
    Ambidextrous
  • Cuff style:
    Knit wrist
  • Liner material:
    Fleece
  • Cat. no.:
    80081-234
  • Supplier No.:
    1040
Specifications
About this item
Clute pattern gloves provide you with general purpose protection at an economical price.
  • Length: 29.5 cm (11⁵/₈")
Split cow leather palm gives you good abrasion protection while the breathable cotton material back keeps your hands cool.