Adjustable Quick-Release Toe Grounding Strap, STATICO
Adjustable Toe Grounder
Catalog # 14223-704
Supplier: Statico
Supplier #: S1140
Jump to:
Product Details & Documents
This Adjustable Toe Grounding Strap provides the end user with an effective means of charge dissipation in ESD sensitive areas
This product is made out of a 2-layer rubber material. Inner layer is beige and insulative, and the outer layer is conductive in black color. Adjustment is made easy through an elastic material and a Quick-Release buckle. The product has a 22" long conductive tab for skin contact and a molded one megohm current limiting safety resistor.
Certifications: Meets ANSI/ESD 20.20-2014.
Specifications
- Description:Adjustable Toe Grounder