Mouse TNFSF18/GITRL PicoKine ELISA Kit, Boster
10205-274
:
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Target protein:TNFSF18/GITRL
- Size:96 well
- Immunogen:Standard: NSO, T47-S173
- Cat. no.:10205-274
- Supplier No.:EK1308
Sandwich ELISA kit of Quantitative Detection for Mouse TNFSF18/GITRL
: Kits include 96-well plate precoated with anti- mouse TNFSF18 antibody 1; ST0000-10 Lyophilized recombinant mouse TNFSF18 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse TNFSF18 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.