Human CD169/SIGLEC-1 PicoKine ELISA Kit, Boster
10205-240
:
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Size:96 well
- Immunogen:Standard: NSO, S20-Q1641
- Cat. no.:10205-240
- Supplier No.:EK1304
Sandwich ELISA kit of Quantitative Detection for Human CD169/SIGLEC-1
: Kits include 96-well plate precoated with anti- human CD169 antibody 1; ST0000-10 Lyophilized recombinant human CD169 standard 10ng/tubex2; AR1107 Biotinylated anti- human CD169 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.