Order Entry
Canada
ContactUsLinkComponent
Mouse ADAM12 PicoKine ELISA Kit, Boster
Mouse ADAM12 PicoKine ELISA Kit, Boster
Catalog # 10207-732
CAS Number:  
undefined
Mouse ADAM12 PicoKine ELISA Kit, Boster
Catalog # 10207-732
Supplier Number:  EK1294
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Size:
    96 well
  • Immunogen:
    Standard: CHO, R29-S513
  • Cat. no.:
    10207-732
  • Supplier No.:
    EK1294

Specifications

About this item

Sandwich ELISA kit of Quantitative Detection for Mouse ADAM12

Ordering information: Kits include 96-well plate precoated with anti- mouse ADAM12 antibody 1; ST0000-10 Lyophilized recombinant mouse ADAM12 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse ADAM12 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.