Order Entry
Canada
ContactUsLinkComponent
Rat Lumican PicoKine ELISA Kit, Boster
Rat Lumican PicoKine ELISA Kit, Boster
Catalog # 10205-676
CAS Number:  
undefined
Rat Lumican PicoKine ELISA Kit, Boster
Catalog # 10205-676
Supplier Number:  EK1262
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Rat
  • Size:
    96 well
  • Immunogen:
    Standard: NSO, Q19-N338
  • Cat. no.:
    10205-676
  • Supplier No.:
    EK1262

Specifications

About this item

Sandwich ELISA kit of Quantitative Detection for Rat Lumican

Ordering information: Kits include 96-well plate precoated with anti- rat Lumican antibody 1; ST0000-10 Lyophilized rat Lumican standard 10ng/tubex2; AR1107 Biotinylated anti- rat Lumican antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.