Order Entry
Canada
ContactUsLinkComponent
Mouse Lumican PicoKine ELISA Kit, Boster
Mouse Lumican PicoKine ELISA Kit, Boster
Catalog # 10205-592
CAS Number:  
undefined
Mouse Lumican PicoKine ELISA Kit, Boster
Catalog # 10205-592
Supplier Number:  EK1261
CAS Number:  

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Size:
    96 well
  • Immunogen:
    Standard: NSO, Q19-N338
  • Cat. no.:
    10205-592
  • Supplier No.:
    EK1261

Specifications

About this item

Sandwich ELISA kit of Quantitative Detection for Mouse Lumican

Ordering information: Kits include 96-well plate precoated with anti- mouse Lumican antibody 1; ST0000-10 Lyophilized recombinant mouse Lumican standard 10ng/tubex2; AR1107 Biotinylated anti- mouse Lumican antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.