Mouse TNFRSF14/HVEM PicoKine ELISA Kit, Boster
10208-772
:
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Size:96 well
- Immunogen:Standard: CHO, Q29-V207
- Cat. no.:10208-772
- Supplier No.:EK1227
Sandwich ELISA kit of Quantitative Detection for Mouse TNFRSF14/HVEM
: Kits include 96-well plate precoated with anti- mouse TNFRSF14 antibody 1; ST0000-10 Lyophilized recombinant mouse TNFRSF14 standard 10ng/tubex2; AR1107 Biotinylated anti- mouse TNFRSF14 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.