Mouse CD13/Aminopeptidase N PicoKine ELISA Kit, Boster
Catalog # 10205-604
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Size:96 well
- Immunogen:Standard: NSO, K69-S966
- Cat. no.:10205-604
- Supplier No.:EK1222
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Mouse CD13/Aminopeptidase N
Ordering information: Kits include 96-well plate precoated with anti- Mouse CD13 antibody 1; ST0000-20 Lyophilized recombinant mouse CD13 standard 20ng/tubex2; AR1107 Biotinylated anti- mouse CD13 antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.