Human SPARC PicoKine ELISA Kit, Boster
Catalog # 10205-668
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Size:96 well
- Immunogen:Standard: NSO, A18-I303
- Cat. no.:10205-668
- Supplier No.:EK1210
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Human SPARC
Ordering information: Kits include 96-well plate precoated with anti- human SPARC antibody 1; ST0000-50 Lyophilized recombinant human SPARC standard 50ng/tubex2; AR1107 Biotinylated anti- human SPARC antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.