Rat IL-17F PicoKine ELISA Kit, Boster
Catalog # 10209-050
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Rat
- Target protein:IL-17F
- Size:96 well
- Immunogen:Standard: E.coli, A19-A153
- Cat. no.:10209-050
- Supplier No.:EK1136
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Rat IL-17F
Ordering information: Kits include 96-well plate precoated with anti- rat IL-17F antibody 1; ST0000-10 Lyophilized recombinant rat IL-17F standard 10ng/tubex2; AR1107 Biotinylated anti- rat IL-17F antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.