Human IL-17RC PicoKine ELISA Kit, Boster
Catalog # 10205-580
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:IL-17C
- Size:96 well
- Immunogen:Standard: NSO, L21-A454
- Cat. no.:10205-580
- Supplier No.:EK0984
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Human IL-17RC
Ordering information: Kits include 96-well plate precoated with anti- human IL-17RC antibody 1; ST0000-10 Lyophilized recombinant human IL-17RC standard 20ng/tubex2; AR1107 Biotinylated anti- human IL-17RC antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.