You searched for: Carboys
Carboys are rigid containers primarily used for transporting liquids, often water or chemicals. In laboratories, the containers are usually made of plastic, though some are still made of ferric glass or other shatter-resistant glasses. With a high chemical resistance and opaque exteriors to block ultraviolet light, the carboys store large quantities of liquids, such as solvents and deionized water, that will remain unaltered. Polypropylene carboys are autoclavable for easy sterilization.
VWR® Carboys with Versatile Caps, HDPE
Supplier: VWR International
VWR® HDPE carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
Expand 12 Items
VWR® Carboys with Versatile Caps, HDPE
Supplier: VWR International
VWR® amber carboys are designed to protect photo-sensitive materials, while providing users with a highly versatile container that maximizes storage efficiency and ease of use.
Expand 3 Items
EZgrip® 20 L Polycarbonate Carboy with 120 mm Cap, Foxx Life Sciences
Supplier: VWR International
The polycarbonate EZgrip® Carboy has molded grips in the base to provide a full and secure grip, and a distinctive rectangular shape to save valuable bench space.
Expand 1 Items
VWR® Carboys with Versatile Caps, PP
Supplier: VWR International
VWR® polypropylene carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
Expand 12 Items
VWR® Copolyester Carboys with Versatile Caps
Supplier: VWR International
VWR® copolyester carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
Expand 4 Items
VWR® Round Carboy, Wide Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 4 Items
VWR® Heavy-Duty Round Carboy, Narrow Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 2 Items
VWR® Round Carboy with Sanitary Neck
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for bioprocessing applications.
Expand 3 Items
VWR® Round Carboy with Sanitary Flange, Narrow Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 2 Items
VWR® Carboy with Closed Cap, Clear PC
Supplier: VWR International
The ergonomically body is coupled with a VersaCap® technology to provide the user an easy-to-use and easy-to-clean carboy.
Expand 1 Items
VWR® Round Carboy, Narrow Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 10 Items
VWR® Carboys with Versatile Caps, Dark Amber PP
Supplier: VWR International
VWR® dark amber carboys are designed to protect photo-sensitive materials, while providing users with a highly versatile container that maximizes storage efficiency and ease of use.
Expand 2 Items
VWR® Rectangular Carboy, Wide Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 3 Items
VWR® Round Carboy with Spigot, Narrow Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 9 Items
VWR® Rectangular Carboy with Spigot, Wide Mouth
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage.
Expand 5 Items
VWR® Amber HDPE Carboys with Spigots
Supplier: VWR International
Smartcap 80. adheres to usp671 standards for light blocking. scratch/chemical resistant. Ideal for mixing powders. handles at top and bottom. open cap and veratile cap adapters provide interchangeable and leakproof top connections.
Expand 3 Items
VWR® Carboys with 3" Sanitary Neck, PP
Supplier: VWR International
VWR® polypropylene carboys are designed to provide users with a highly versatile container that maximizes storage efficiency and ease of use.
Expand 1 Items
VWR® Carboy with Sanitary Neck
Supplier: VWR International
Sterile carboy with sanitary neck, made from HDPE.
Expand 1 Items
VWR® Round Carboy, Sterile
Supplier: VWR International
Round, sterile carboy made from HDPE.
Expand 1 Items
VWR® EZLabpure™ Round Carboys, with 83B VersaCap®
Supplier: VWR International
VWR® EZLabpure™ round carboys are manufactured using high-purity virgin USP Class VI medical grade polypropylene.
Expand 7 Items
VWR® Round Carboy, Narrow Mouth, Sterile
Supplier: VWR International
Use for storing and dispensing solutions and media. Ideal for sterile water storage. These high-density polyethylene, disposable carboys are ideal for storing and transfer of sterile fluids and pharmaceutical/biotech reagents.
Expand 2 Items
EZBio® Bioprocess, Single-Use, Double Bagged, Sterile Carboys, Foxx Life Sciences
Supplier: VWR International
EZBio® single use carboys are available in amber HDPE, dark amber polypropylene, HDPE, polyethylene terephthalate glycol-modified (PETG), and polycarbonate (PC).
Expand 29 Items
Nalgene® Rectangular Carboys with Handle, High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sturdy, space-saving design with molded-in graduations in liters and gallons.
Expand 2 Items
WHEATON® Jugs, HDPE, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Manufactured from high density polyethylene.
Expand 1 Items
SP Bel-Art Carboy with Handle and Screw Cap, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
This space-saving sturdy polyethylene carboy has an easy-grip handle and screw cap for leakproof shipping and storage.
Expand 1 Items
Round Carboys
Supplier: Thermo Fisher Scientific
Suitable for collecting and dispensing distilled water, reagents, and acids, these low-density polyethylene carboys are graduated in 1 gallon and 5 L increments.
Expand 3 Items
Nalgene® Certified Platinum Clean HDPE Bottles and Carboys, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Thermo Scientific Nalgene® certified platinum clean HDPE bottles and carboys help reduce the potential for carry-over contamination.
Expand 10 Items
Cubitainer Containers, Low-Density Polyethylene, Qorpak®
Supplier: Qorpak
Cubitainers® are low-density flexible polyethylene cube-shaped inserts inside a heavy-duty corrugated outer carton (32 ounce does not include carton).
Expand 1 Items
Nalgene® Lowboy™ Carboys with Spigot, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Carboys are ideal for storing and dispensing stock solutions.
Expand 1 Items
Nalgene® Carboys with Handles, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Excellent for storing culture media, distilled water, and other solutions.