Order Entry
Canada
ContactUsLinkComponent
Human KIT/SCFR PicoKine ELISA Kit, Boster
Human KIT/SCFR PicoKine ELISA Kit, Boster
  10207-676
 :  
undefined
Human KIT/SCFR PicoKine ELISA Kit, Boster
  10207-676
 :  EK0835
 :  

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Size:
    96 well
  • Immunogen:
    Standard: sf21, Q26-T520
  • Cat. no.:
    10207-676
  • Supplier No.:
    EK0835

 

 

Sandwich ELISA kit of Quantitative Detection for Human KIT/SCFR

 : Kits include 96-well plate precoated with anti- human KIT/SCFR antibody 1; ST0000-50 Lyophilized recombinant human KIT/SCFR standard 50ng/tubex2; AR1107 Biotinylated anti- human KIT/SCFR antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.