Order Entry
Canada
ContactUsLinkComponent
Mouse MER/MERTK PicoKine ELISA Kit, Boster
Mouse MER/MERTK PicoKine ELISA Kit, Boster
Catalog # 10205-486
CAS Number:  
undefined
Mouse MER/MERTK PicoKine ELISA Kit, Boster
Catalog # 10205-486
Supplier Number:  EK0811
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Size:
    96 well
  • Immunogen:
    Standard: sf21, E23-F498
  • Cat. no.:
    10205-486
  • Supplier No.:
    EK0811

Specifications

About this item

Sandwich ELISA kit of Quantitative Detection for Mouse MER/MERTK

Ordering information: Kits include 96-well plate precoated with anti- mouse MER antibody 1; ST0000-10 Lyophilized recombinant mouse MER standard 10ng/tubex2; AR1107 Biotinylated anti- mouse MER antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.