Order Entry
Canada
ContactUsLinkComponent
Human CSF1R/M-CSFR PicoKine ELISA Kit, Boster
Human CSF1R/M-CSFR PicoKine ELISA Kit, Boster
  10207-590
 :  
undefined
Human CSF1R/M-CSFR PicoKine ELISA Kit, Boster
  10207-590
 :  EK0807
 :  

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Size:
    96 well
  • Immunogen:
    Standard: NSO, I20-E512
  • Cat. no.:
    10207-590
  • Supplier No.:
    EK0807

 

 

Sandwich ELISA kit of Quantitative Detection for Human CSF1R/M-CSFR

 : Kits include 96-well plate precoated with anti- human CSF1R/M-CSFR antibody 1; ST0000-10 Lyophilized recombinant human CSF1R/M-CSFR standard 10ng/tubex2; AR1107 Biotinylated anti- human CSF1R/M-CSFR antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.