Order Entry
ContactUsLinkComponent
Human IL-17C PicoKine ELISA Kit, Boster
Human IL-17C PicoKine ELISA Kit, Boster
Catalog # 10209-920
CAS Number:  
undefined
Human IL-17C PicoKine ELISA Kit, Boster
Catalog # 10209-920
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-17C
  • Size:
    96 well
  • Immunogen:
    Standard: E.coli, H19-V197
  • Cat. no.:
    10209-920
  • Supplier No.:
    EK0789

Specifications

About this item

Sandwich ELISA kit of Quantitative Detection for Human IL-17C

Ordering information: Kits include 96-well plate precoated with anti- human IL-17C antibody 1; ST0000-10 Lyophilized recombinant human IL-17C standard 10ng/tubex2; AR1107 Biotinylated anti- human IL-17C antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.
Frequently Bought Together