Human IGF1R PicoKine ELISA Kit, Boster
10207-832
:
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Size:96 well
- Immunogen:Standard: NSO, E31-N932
- Cat. no.:10207-832
- Supplier No.:EK0776
Sandwich ELISA kit of Quantitative Detection for Human IGF1R
: Kits include 96-well plate precoated with anti- human IGF1R antibody 1; ST0000-10 Lyophilized recombinant human IGF1R standard 10ng/tubex2; AR1107 Biotinylated anti- human IGF1Rantibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex (ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30 ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.