Human VEGFR2/KDR PicoKine ELISA Kit, Boster
10205-358
:
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Target protein:sVEGF-R2
- Size:96 well
- Immunogen:Standard: NSO, A20-E764
- Cat. no.:10205-358
- Supplier No.:EK0544
Sandwich ELISA kit of Quantitative Detection for Human VEGFR2/KDR
: Kits include 96-well plate precoated with anti- human VEGFR2/KDR antibody 1; ST0000-10 Lyophilized recombinant human VEGFR2/KDR standard 10ng/tubex2; AR1107 Biotinylated anti- human VEGFR2/KDR antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.