Human uPAR PicoKine ELISA Kit, Boster
10207-806
:
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Size:96 well
- Immunogen:Standard: NSO,L23-R303
- Cat. no.:10207-806
- Supplier No.:EK0536
Sandwich ELISA kit of Quantitative Detection for Human uPAR
: Kits include 96-well plate precoated with anti- human uPAR antibody 1; ST0000-10 Lyophilized recombinant human uPAR standard 10ng/tubex2; AR1107 Biotinylated anti- human uPAR antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.