Human uPA/PLAU PicoKine ELISA Kit, Boster
Catalog # 10205-530
Supplier: Boster Biological Technology
CAS Number:
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:Human
- Size:96 well
- Immunogen:Standard: NSO, M1-L431
- Cat. no.:10205-530
- Supplier No.:EK0535
Specifications
About this item
Sandwich ELISA kit of Quantitative Detection for Human uPA/PLAU
Ordering information: Kits include 96-well plate precoated with anti- human uPA/PLAU antibody 1; ST0000-10 Lyophilized recombinant human uPA/PLAU standard 10ng/tubex2; AR1107 Biotinylated anti- human uPA/PLAU antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.