Order Entry
Canada
ContactUsLinkComponent
 
Human OPN PicoKine ELISA Kit, Boster
  10207-892
 :  
undefined
Human OPN PicoKine ELISA Kit, Boster
  10207-892
 :  EK0482
 :  

 

  • Assay Type:
    Sandwich
  • Primary antibody reactivity:
    Human
  • Size:
    96 well
  • Immunogen:
    Standard: NSO, I17-N300
  • Cat. no.:
    10207-892
  • Supplier No.:
    EK0482

 

 

Sandwich ELISA kit of Quantitative Detection for Human OPN

 : Kits include 96-well plate precoated with anti- human OPN antibody 1; ST0000-10 Lyophilized recombinant human OPN standard 10ng/tubex2; AR1107 Biotinylated anti- human OPN antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.