Mouse NGF/NGF beta PicoKine ELISA Kit, Boster
10205-278
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Size:96 well
- Immunogen:Standard: NSO, S122-G241
- Cat. no.:10205-278
- Supplier No.:EK0470
Sandwich ELISA kit of Quantitative Detection for Mouse NGF/NGF beta
: Kits include 96-well plate precoated with anti- mouse NGF/NGF beta antibody 1; ST0000-10 Lyophilized recombinant mouse NGF/NGF beta standard 10ng/tubex2; AR1107 Biotinylated anti- mouse NGF/NGF beta antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.