Mouse GM-CSF PicoKine ELISA Kit, Boster
10209-016
:
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Target protein:GM-CSF
- Size:96 well
- Immunogen:Standard: E.coli, A18-K141
- Cat. no.:10209-016
- Supplier No.:EK0365
Sandwich ELISA kit of Quantitative Detection for Mouse GM-CSF
: Kits include 96-well plate precoated with anti- mouse GM-CSF antibody 1; ST0000-10 Lyophilized recombinant mouse GM-CSF standard 10ng/tubex2; AR1107 Biotinylated anti- mouse GM-CSF antibody 130ul(dilution 1:100); AR1103 Avidin-Biotin-Peroxidase Complex(ABC) 130ul(dilution 1:100); AR1106-1 Sample diluent buffer 30ml; AR1106-2 Antibody diluent buffer 12ml; AR1106-3 ABC diluent buffer 12ml; AR1104 TMB color developing agent 10ml; AR1105 TMB stop solution 10ml.