You searched for: Peristaltic Pumps
Peristaltic pumps rely on compressing and releasing a length of flexible tubing to mimic natural peristalsis seen in many biological systems, using positive displacement to move fluids in a pulsing manner. Procedures will remain contamination-free using these systems, since the only part of the pump touched by the solution is the interior of the tubing. Additionally, the pumps are able to handle heavier and more aggressive fluids. The flexible tubing requires few periodic replacement and maintenance needs.
Masterflex® Peristaltic Injector Pumps, Avantor®
Supplier: VWR International
A cost-effective pump for pressure injection and chemical feed applications.
Expand 5 Items
Masterflex® L/S® Digital Process Pump Systems, Avantor®
Supplier: VWR International
Transfer, dispense, and meter in lab, process and washdown environments.
Expand 6 Items
Masterflex® I/P® Digital Network-Compatible Pump Systems, Avantor®
Supplier: VWR International
Networking drives offer either native EtherNet/IP™ or Profibus communication—no additional hardware or firmware required.
Expand 44 Items
Masterflex® High-Pressure Peristaltic Pumps, Low-Flow Range, Avantor®
Supplier: VWR International
Don’t let back pressure affect output.
Expand 1 Items
Masterflex® Ismatec® IP and IP-N Digital Multichannel Peristaltic Pumps, Avantor®
Supplier: VWR International
Smooth, precise flow and fewer tubing changes.
Expand 35 Items
Masterflex® I/P® MasterSense™ Pump Systems
Supplier: VWR International
Versatile pumps for process applications – with an intuitive touch-screen interface and advanced connectivity.
Expand 18 Items
Masterflex® L/S® MasterSense™ Fill/Finish Pump System with Masterflex Ultrapharm™ Pump Head, Avantor®
Supplier: VWR International
A highly accurate benchtop pump system for dosing, dispensing, and fill/finish applications.
Expand 18 Items
Masterflex® Micro Peristaltic Pumps, Avantor®
Supplier: VWR International
Operate by computer or on your iOS or Android™ device.
Expand 4 Items
Masterflex® I/P® Digital Precision Modular Drive Systems, Avantor®
Supplier: VWR International
Place motor and controller where convenient with modular design.
Expand 3 Items
Masterflex® L/S® Standard Digital Pump Systems, Avantor®
Supplier: VWR International
Designed with maintenance-free high-accuracy motor and a graphical interface—makes setup and operation easier than ever!
Expand 8 Items
Masterflex® L/S® MasterSense™ Pump Systems
Supplier: VWR International
Complete pump systems for accurate fluid delivery with an intuitive touch-screen interface.
Expand 36 Items
Masterflex® B/T® MasterSense™ BioPharma Process Pump, Avantor®
Supplier: VWR International
High-flow pump for process applications – with an intuitive touch-screen interface and advanced connectivity.
Expand 2 Items
Masterflex® Modular Touch-Screen Dispensing Pump, Avantor®
Supplier: VWR International
A versatile dispensing pump with an intuitive touch-screen interface for programming and control.
Expand 3 Items
Masterflex® Ismatec® Reglo Independent Channel Control (ICC) Peristaltic Pumps, Avantor®
Supplier: VWR International
Independently control flow and direction of each channel via keypad or your PC.
Expand 10 Items
Masterflex® L/S® Digital Process Pump Systems with Cytoflow™ Pump Head, Avantor®
Supplier: Avantor Fluid Handling
Ideal for the transfer and dispensing of live cell cultures, media, and other shear-sensitive fluids in process and washdown environments.
Expand 2 Items
Masterflex® L/S® Network-Compatible Pump Systems, Avantor®
Supplier: VWR International
Networking drives offer either native EtherNet/IP™ or Profibus communication—no additional hardware or firmware required.
Expand 12 Items
Masterflex® L/S® Precision Process Pump Systems, Avantor®
Supplier: VWR International
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
Expand 8 Items
Masterflex® B/T® Fixed Speed Analog Drives, Avantor®
Supplier: Avantor Fluid Handling
High-volume transfer and dispensing.
Expand 2 Items
Masterflex® L/S® Precision Variable-Speed Modular Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
Place where convenient—ideal for limited spaces, hoods, or isolation chambers.
Expand 3 Items
Masterflex® Ismatec® Fixed-Speed Rack-Mount Minicartridge Peristaltic Pumps, Avantor®
Supplier: Avantor Fluid Handling
Compact size mounting rod—use these pumps almost anywhere.
Expand 19 Items
Masterflex® L/S® Analog Variable-Speed Console Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
Compact, cost effective pump systems for general fluid transfer applications.
Expand 7 Items
Masterflex® I/P® Precision Brushless Process Pumps, Avantor®
Supplier: Avantor Fluid Handling
Powerful, low-maintenance drives are versatile and cost-effective.
Expand 18 Items
Masterflex® C/L® Microflex® Digital Pump System, Avantor®
Supplier: Avantor Fluid Handling
Small footprint digital drive delivers accurate low flow over a wide range.
Expand 1 Items
Masterflex® L/S Digital Dispensing Systems with Easy Load II Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
Precision digital drive offers a maintenance-free high-accuracy motor and a graphical interface – makes setup and operation easier than ever.
Expand 6 Items
Masterflex® L/S® High Pressure Precision Process Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
Pump at pressures up to 125 psi (8.5 bar).
Expand 3 Items
Masterflex® I/P® Stainless-Steel IP66 Digital Dispensing Pump System, Avantor®
Supplier: Avantor Fluid Handling
Dispense by Volume in mL (0.001 to 99999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59 min, 59 sec per cycle).
Expand 1 Items
Masterflex® L/S® Standard Digital PTFE-Tubing Pump System, Avantor®
Supplier: Avantor Fluid Handling
Transfer aggressive organic solvents and high-purity fluids.
Expand 1 Items
Masterflex® B/T® Variable Speed and Air-Powered Analog Drives, Avantor®
Supplier: Avantor Fluid Handling
High-volume transfer and dispensing.
Expand 4 Items
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
Supplier: Avantor Fluid Handling
Versatile pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
Expand 2 Items
Masterflex® L/S® Variable Speed Analog Console Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
Don’t lose control of your metering applications—digital display gives you precise control.