HSP70 ELISA Kit, Enzo Life Sciences
Catalog # 89141-858
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Target protein:HSP70
- Sensitivity:200 pg/ml (range 780 - 50,000 pg/ml)
- Cat. no.:89141-858
- Supplier No.:EKS-700B
- No. of Tests:96 wells
Specifications
About this item
Widely used ELISA HSP70 kit for chaperone and stress research.
For the quantitative determination of HSP70 in cell lysates and tissue of human, mouse, and rat origin. Prod. No. ADI-EKS-715 is recommended for the detection of Hsp70 in serum and plasma samples.
The HSP70 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4.5 hours. Absorbance is read at 450 nm. This kit is ideal for measuring intracellular HSP70 (HSP72). Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Ordering information: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent 2, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.