Specifications
- Size:1 Library
- Cat. no.:89156-286
- Supplier No.:2820-0500
Specifications
About this item
The Screen-Well Purinergic and Adenosine Ligands Plate contains 72 CNS receptor ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Purinergic and Adenosine Ligands Plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.