ContactUsLinkComponent
SCREEN-WELL® Metabotropic Glutamatergic Ligand Library, Enzo Life Sciences
SCREEN-WELL® Metabotropic Glutamatergic Ligand Library, Enzo Life Sciences
  89156-282
 :  
SCREEN-WELL® Metabotropic Glutamatergic Ligand Library, Enzo Life Sciences
  89156-282
 :  BML-2818-0500
 :  
  • Size:
    1 Library
  • Cat. no.:
    89156-282
  • Supplier No.:
    2818-0500
The Metabotropic Glutamatergic Ligands Plate contains 49 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs.
The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Metabotropic Glutamatergic Ligands plate is 1 plate of the larger Neurotransmitter Library which contains over 650 ligands.